missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92806-0.1ml
This item is not returnable.
View return policy
Description
ATG9B Polyclonal antibody specifically detects ATG9B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ATG9B | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| APG9L2APG9-like 2, ATG9 autophagy related 9 homolog B (S. cerevisiae), autophagy-related protein 9B, endothelial nitric oxide synthase antisense, nitric oxide synthase 3 antisense, Nitric oxide synthase 3-overlapping antisense gene protein, NOS3ASautophagy 9-like 2 protein, Protein sONE, SONEFLJ14885 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 715-924 of human ATG9B (NP_001303985.1). HPLWRPPGHSSKFLGHLWGRVQQDAAAWGATSARGPSTPGVLSNCTSPLPEAFLANLFVHPLLPPRDLSPTAPCPAAATASLLASISRIAQDPSSVSPGGTGGQKLAQLPELASAEMSLHVIYLHQLHQQQQQQEPWGEAAASILSRPCSSPSQPPSPDEEKPSWSSDGSSPASSPRQQWGTQKARNLFPGGFQVTTDTQKEPDRASCTD | |
| 0.1 mL | |
| Autophagy, Golgi Apparatus Markers | |
| 285973 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction