missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG9B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | ATG9B |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668921
|
Novus Biologicals
NBP2-92806-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627772
|
Novus Biologicals
NBP2-92806-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG9B Polyclonal antibody specifically detects ATG9B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ATG9B | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Golgi Apparatus Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 285973 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG9L2APG9-like 2, ATG9 autophagy related 9 homolog B (S. cerevisiae), autophagy-related protein 9B, endothelial nitric oxide synthase antisense, nitric oxide synthase 3 antisense, Nitric oxide synthase 3-overlapping antisense gene protein, NOS3ASautophagy 9-like 2 protein, Protein sONE, SONEFLJ14885 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 715-924 of human ATG9B (NP_001303985.1). HPLWRPPGHSSKFLGHLWGRVQQDAAAWGATSARGPSTPGVLSNCTSPLPEAFLANLFVHPLLPPRDLSPTAPCPAAATASLLASISRIAQDPSSVSPGGTGGQKLAQLPELASAEMSLHVIYLHQLHQQQQQQEPWGEAAASILSRPCSSPSQPPSPDEEKPSWSSDGSSPASSPRQQWGTQKARNLFPGGFQVTTDTQKEPDRASCTD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title