Learn More
ATL3 Mouse, anti-Human, Clone: 2E7C4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68516-1-IG-20UL
 View more versions of this product
Product Code. 18066165
  • € 145.00 / 20µL
Description
The ATL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATL3. This antibody recognizes human, mouse, rat, rabbit antigen. The ATL3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ATL3
2E7C4
ATL3, Atlastin 3, atlastin GTPase 3, DKFZp564J0863
20 μL
25923
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Rabbit
150 kDa