missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATL3 Mouse, anti-Human, Clone: 2E7C4, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ATL3 |
|---|---|
| Clone | 2E7C4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18088695
|
Proteintech
68516-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18066165
|
Proteintech
68516-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ATL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATL3. This antibody recognizes human, mouse, rat, rabbit antigen. The ATL3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ATL3 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Rabbit | |
| ATL3, Atlastin 3, atlastin GTPase 3, DKFZp564J0863 | |
| Primary |
| 2E7C4 | |
| Western Blot, ELISA | |
| Mouse | |
| 25923 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit