Learn More
ATP6V1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14780-1-AP-20UL
 View more versions of this product
Product Code. 16843594
  • € 145.00 / 20µL
Description
Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, while F-ATPases reverse the process, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and a integral V0 domain, which is responsible for proton translocation, compose V-ATPase. Nine subunits (A-H) make up the V1 domain and five subunits (a, d, c, c′ and c") make up the V0 domain. Like F-ATPase, V-ATPase most likely operates through a rotary mechanism. The V-ATPase V1 B subunit exists as two isoforms. In the inner ear, the V-ATPase B1 isoform functions in proton secretion and is required to maintain proper endolymph pH and normal auditory function. The gene encoding the human V-ATPase B1 isoform maps to chromosome 2cen-q13. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. The V-ATPase B2 isoform is expressed in kidney and is the only B isoform expressed in osteoclasts.
Specifications
Specifications
ATP6V1B1
Polyclonal
Unconjugated
ATP6V1B1
ATP6B1, ATP6V1B1, RTA1B, V ATPase subunit B 1, VATB, VMA2, VPP3
Rabbit
Antigen Affinity Chromatography
RUO
110935, 312488, 525
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15313
ATP6V1B1
ATP6V1B1 Fusion Protein Ag6332
20 μL
Primary
Mouse, Human, Rat