missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aurora A/B Monoclonal Antibody (3H1)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547320

 View more versions of this product

Product Code. 17825257

  • € 1750.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Aurora A/B Monoclonal Antibody for Western Blot, ICC/IF, IHC

The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aurora A/B
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5 mM sodium azide
O14965, O55099, O70126, P59241, P97477, Q96GD4
Mouse
Protein A/G
RUO
114592, 20877, 20878, 261730, 6790, 9212
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot, Immunocytochemistry
3H1
Unconjugated
AURKA
AURKA, AURKB
Full length recombinant human Aurora A protein expressed in and purified from E. coli.
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.