missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aurora C Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 208.00 - € 476.00
Specifications
| Antigen | Aurora C |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17760384
![]() |
Invitrogen™
PA5118973 |
200 μL |
€ 476.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17783614
|
Invitrogen™
PA5122220 |
20 μL |
€ 208.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aurora C Polyclonal Antibody for Western Blot, IHC (P)
AURKC (Aurora C) is a chromosomal passenger protein that forms complexes with AURKB (Aurora B) and inner centromere proteins, playing a role in organizing microtubules in relation to centrosome/spindle function during mitosis. It is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.Specifications
| Aurora C | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| AURKC | |
| AIE1; AIE2; AIK3; AIRK3; ARK3; ARK-3; AurC; AURKC; Aurora 3; aurora B; aurora kinase C; aurora/Ipl1/Eg2 protein 1; aurora/IPL1/EG2 protein 2; aurora/IPL1-related kinase 3; aurora-C; aurora-related kinase 3; epididymis secretory protein Li 90; HEL-S-90; IAK3; serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine kinase 13 (aurora/IPL-like); serine/threonine-protein kinase 13; Serine/threonine-protein kinase aurora-C; SPGF5; STK13 | |
| AURKC | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O88445 | |
| 20871 | |
| Recombinant protein Aurora Kinase C. The antigen corresponds to amino acid range 5-237 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title