Learn More
Invitrogen™ AVEN Recombinant Rabbit Monoclonal Antibody (24GB6865)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556989
 View more versions of this product
Product Code. 30276363
  • € 500.00 / 100µL
Description
AVEN Recombinant Monoclonal Antibody for Western Blot
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Aven is a recently discovered protein that blocks apoptosis induced by Apaf-1 and caspase-9. It is thought that Aven functions by binding to Bcl-xL, an antiapoptotic member of the Bcl-2 family, and to Apaf-1, possibly interfering with the ability of Apaf-1 to self-associate, suggesting that Aven impedes Apaf-1-mediated caspase activation. Higher levels of Aven mRNA are seen in patients with acute leukemia than in control patients, suggesting that Aven may be useful as a prognostic indicator in leukemia patients.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AVEN
Recombinant Monoclonal
Unconjugated
AVEN
1700013A01Rik; 1700056A21Rik; apoptosis and caspase activation inhibitor; apoptosis, caspase activation inhibitor; AVEN; Cell death regulator Aven; mAven-L; mAven-S; PDCD12; programmed cell death 12
Rabbit
Affinity chromatography
RUO
311299, 57099, 74268
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6865
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9D9K3, Q9NQS1
AVEN
A synthesized peptide derived from human AVEN (aa 113-126).
100 μL
Primary
Human, Mouse, Rat