missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-41066
This item is not returnable.
View return policy
Description
Avian Influenza Hemagglutinin 4 Polyclonal specifically detects Avian Influenza Hemagglutinin 4 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Avian Influenza Hemagglutinin 4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide corresponding to 10 amino acids near the center of the Hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 260 - 310 of Avian Influenza Hemagglutinin 4. Amino Acid Squence: REDIMKSELEYGN | |
| 0.1 mg | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot, ELISA 1 μg/mL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction