missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
€ 178.00 - € 507.00
Specifications
| Antigen | Avian Influenza Hemagglutinin 4 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690696
|
Novus Biologicals
NBP2-41066-0.025mg |
0.025 mg |
€ 178.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18171169
|
Novus Biologicals
NBP2-41066 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Avian Influenza Hemagglutinin 4 Polyclonal specifically detects Avian Influenza Hemagglutinin 4 in Virus samples. It is validated for Western Blot, ELISA.Specifications
| Avian Influenza Hemagglutinin 4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Polyclonal | |
| Goat | |
| Virus | |
| Antibody was raised against a synthetic peptide corresponding to 10 amino acids near the center of the Hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 260 - 310 of Avian Influenza Hemagglutinin 4. Amino Acid Squence: REDIMKSELEYGN | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title