Learn More
Invitrogen™ AZI2 Recombinant Rabbit Monoclonal Antibody (24GB3555)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556948
This item is not returnable.
View return policy
Description
AZI2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).
Specifications
| AZI2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AZI2 | |
| 5-azacytidine induced 2; 5-azacytidine induced gene 2; 5-azacytidine-induced protein 2; AA410145; Az2; AZI2; Nak-associated protein 1; NAP1; NF-kappa-B-activating kinase-associated protein 1; TBKBP2; Tbkp2; TILP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27215, 316051, 64343 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3555 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4KMA0, Q9H6S1, Q9QYP6 | |
| AZI2 | |
| A synthesized peptide derived from human AZI2 (aa 280-392). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |