missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AZI2 Recombinant Rabbit Monoclonal Antibody (24GB3555)
Rabbit Recombinant Monoclonal Antibody
€ 146.00 - € 500.00
Specifications
| Antigen | AZI2 |
|---|---|
| Clone | 24GB3555 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30275966
|
Invitrogen™
MA556947 |
100 μL |
€ 500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276473
|
Invitrogen™
MA556948 |
20 μL |
€ 146.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AZI2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).Specifications
| AZI2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| AZI2 | |
| 5-azacytidine induced 2; 5-azacytidine induced gene 2; 5-azacytidine-induced protein 2; AA410145; Az2; AZI2; Nak-associated protein 1; NAP1; NF-kappa-B-activating kinase-associated protein 1; TBKBP2; Tbkp2; TILP | |
| AZI2 | |
| Primary | |
| Affinity Chromatography |
| 24GB3555 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4KMA0, Q9H6S1, Q9QYP6 | |
| 27215, 316051, 64343 | |
| A synthesized peptide derived from human AZI2 (aa 280-392). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title