missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Raf, p Ser445 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05377-100ul
This item is not returnable.
View return policy
Description
B-Raf Polyclonal antibody specifically detects B-Raf in Human, Rat samples. It is validated for Western Blot
Specifications
| B-Raf | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 | |
| A synthetic phosphorylated peptide around S445 of human BRAF (NP_004324.2). RDSSD | |
| 100 μg | |
| Apoptosis, Phospho Specific | |
| 673 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction