missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Raf, p Ser445 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 479.00
Specifications
| Antigen | B-Raf |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624669
|
Novus Biologicals
NBP3-05377-100ul |
100 μg |
€ 479.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673108
|
Novus Biologicals
NBP3-05377-20ul |
20 μg |
€ 201.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B-Raf Polyclonal antibody specifically detects B-Raf in Human, Rat samples. It is validated for Western BlotSpecifications
| B-Raf | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Phospho Specific | |
| PBS with 50% glycerol, pH7.3. | |
| 673 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 | |
| A synthetic phosphorylated peptide around S445 of human BRAF (NP_004324.2). RDSSD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title