missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ B7-H3 (CD276) Recombinant Rabbit Monoclonal Antibody (BLR026F)

Rabbit Recombinant Monoclonal Antibody
€ 148.00 - € 452.00
Specifications
| Antigen | B7-H3 (CD276) |
|---|---|
| Clone | BLR026F |
| Concentration | 1 mg/mL |
| Content And Storage | 4°C |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17735619
![]() |
Invitrogen™
MA544247 |
100 μL |
€ 452.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17745619
|
Invitrogen™
MA544248 |
10 μL |
€ 148.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.Specifications
| B7-H3 (CD276) | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.09% sodium azide; pH 8.2 | |
| Q5ZPR3 | |
| 80381 | |
| Recombinant protein representing the extracellular domain of human CD276 (residues 29-465). | |
| Antibody |
| BLR026F | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| CD276 | |
| 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 | |
| CD276 | |
| Primary | |
| purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title