missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAP1 Antibody (BAP1/9795R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-44624-100ug
This item is not returnable.
View return policy
Description
BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry
Specifications
| BAP1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| BAP1/9795R | |
| Unconjugated | |
| BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2 | |
| Recombinant fragment (around aa400-729) of human BAP1 protein (Uniprot: Q92560). Exact sequence is proprietary. | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors | |
| 8314 | |
| Store at 4⪚C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction