missing translation for 'onlineSavingsMsg'
Learn More

BAP1 Antibody (BAP1/9795R), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 289.00 - € 518.00

Specifications

Antigen BAP1
Clone BAP1/9795R
Concentration 0.2 mg/ml
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30288137
View Documents
Novus Biologicals
NBP3-44624-20ug
20 μg
€ 289.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30293741
View Documents
Novus Biologicals
NBP3-44624-100ug
100 μg
€ 518.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

BAP1
0.2 mg/ml
Monoclonal
Purified
RUO
Human
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Recombinant fragment (around aa400-729) of human BAP1 protein (Uniprot: Q92560). Exact sequence is proprietary.
Primary
Store at 4⪚C.
BAP1/9795R
Immunohistochemistry
Unconjugated
Rabbit
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
10 mM PBS with 0.05% BSA
8314
IgG κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.