Learn More
BBS5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14569-1-AP-150UL
 View more versions of this product
Product Code. 16893264
  • € 410.00 / 150µL
Description
The BBS5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BBS5. This antibody recognizes human, mouse, rat antigen. The BBS5 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
BBS5
Polyclonal
Unconjugated
150 μL
129880
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
300 μg/mL
Rabbit
Primary
Human, Rat, Mouse