missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | BBS5 |
|---|---|
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Primary or Secondary | Primary |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16803274
|
Proteintech
14569-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16893264
|
Proteintech
14569-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BBS5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BBS5. This antibody recognizes human, mouse, rat antigen. The BBS5 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| BBS5 | |
| Unconjugated | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| Rabbit | |
| Polyclonal |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title