missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Mouse anti-Mouse, Rat, Clone: 2H12, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11881-100UG
This item is not returnable.
View return policy
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Mouse, Rat samples. It is validated for Western Blot
Specifications
| bcl-x | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| A synthetic peptide corresponding to amino acids 3 to 14 of the human bcl-xl sequence (Uniprot: Q07817) | |
| RUO | |
| 598 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 2H12 | |
| Western Blot 1 - 5 ug/ml | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction