missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Mouse anti-Mouse, Rat, Clone: 2H12, Novus Biologicals™
Mouse Monoclonal Antibody
€ 224.00 - € 530.00
Specifications
| Antigen | bcl-x |
|---|---|
| Clone | 2H12 |
| Dilution | Western Blot 1 - 5 ug/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388361
|
Bio-Techne
NBP3-11881-25UG |
25 μg |
€ 224.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18014764
|
Novus Biologicals
NBP3-11881-100UG |
100 μg |
€ 530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Mouse, Rat samples. It is validated for Western BlotSpecifications
| bcl-x | |
| Western Blot 1 - 5 ug/ml | |
| Monoclonal | |
| Mouse | |
| Mouse, Rat | |
| 598 | |
| IgG2a | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 2H12 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS | |
| A synthetic peptide corresponding to amino acids 3 to 14 of the human bcl-xl sequence (Uniprot: Q07817) | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title