Learn More
BD Difco™ Dehydrated Culture Media: Brilliant Green Bile 2%
Standard Methods medium for confirmed and completed tests for coliform bacteria in water and foods
Brand: BD 271000
Additional Details : Weight : 0.80000kg
Specifications
AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW | |
Dehydrated | |
Microbiology Media | |
Standard method medium for confirmed and completed tests for Coliform bacteria in Water and foods | |
2 kg |
2°C to 25°C | |
Brilliant Green Bile 2% | |
Peptone 10g/L, Oxgall 20g/L, Lactose 10g/L, Brilliant green 13.3mg/L | |
Pail |