Learn More
BDH1 Mouse anti-Human, Unconjugated, Clone: 2C3D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67448-1-IG-150UL
Description
The BDH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BDH1. This antibody recognizes human, mouse, rat, pig antigen. The BDH1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| BDH1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2C3D4 | |
| Unconjugated | |
| 150 μL | |
| 622 | |
| IgG1 |