missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDH1 Mouse anti-Human, Unconjugated, Clone: 2C3D4, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | BDH1 |
|---|---|
| Clone | 2C3D4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16820086
|
Proteintech
67448-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16810086
|
Proteintech
67448-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BDH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BDH1. This antibody recognizes human, mouse, rat, pig antigen. The BDH1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| BDH1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 622 | |
| Primary |
| 2C3D4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Mouse, Rat, Pig | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title