Learn More
Bel Art™ Lab Companion Cabinet Style Vacuum Desiccator, Clear
Product Code. 15766879 Shop All SP Scienceware Products
Click to view available options
Volume (Metric):
11 L
23 L
35 L
45 L
Unit Size:
Each
Product Code. 15766879
Brand: Bel Art™ F424004031
Only null left
Add to basket
Add to basket
Includes
Desiccant tray (desiccants sold separately)
Clear desiccator provides vacuum sustainment in a space maximizing cube.
  • Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods
  • Viton™ 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange
  • Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents
  • Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples
  • Use with any vacuum with free air capacity of 1.5 cubic feet per minute (CFM) ft or greater
  • Single piece molded design (except the door) enables outstanding vacuum capability with virtually no leakage and exceptional durability
  • Cabinet style provides maximum useable space
  • Hinged door allows easy access; built in locking tabs secure the door when not under vacuum
  • Analog vacuum gauge in front door allows vacuum level checking at a glance
  • Perforated shelves can be repositioned to accommodate various storage needs (additional shelves sold separately)
  • Built-in ports permit attachment of up to 3 additional valves (sold separately)
  • Stackable space-saving design allows efficient use of work space
  • Eight models to choose from; 11, 23, 35, and 45 liter internal volume in clear or UV blocking amber
Specifications
Color Clear
Material Polycarbonate
Product Type Vacuum Desiccator
Door Type Hinged
Includes Desiccant tray, 3 perforated shelves, built-in ports, door-mounted analog vacuum gauge
Max. Shelf Load 5 kg
No. of Shelves 3/6
Style Cabinet
Volume (English) 1.6 cu. ft.
Volume (Metric) 45 L
Dimensions (D x W x H) Interior 14.75 x 14 x 17.5 in. (37.46 x 35.56 x 44.5 cm)
Width (English) 16.5 in.
Width (English) Door 11.6 in.
Width (English) Interior 14 in.
Width (Metric) 42 cm
Width (Metric) Door 29.5 cm
Width (Metric) Interior 35.56 cm
Height (Metric) 49.1 cm
Height (English) 19.37 in.
Height (English) Door Opening 17.5 in.
Height (English) Interior 17.5 in.
Height (Metric) Door Opening 44.5 cm
Height (Metric) Interior 44.5 cm
Depth (English) 15.62 in.
Depth (English) Interior 14.75 in.
Depth (Metric) 39.7 cm
Depth (Metric) Interior 37.46 cm
Diameter (English) Outer 15.62 in.
Diameter (Metric) Inner 42 cm
Diameter (Metric) Outer 39.67 cm
Weight (English) 26.75 lb.
Weight (Metric) 12.2 kg