Learn More
beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47700B-100ul
 View more versions of this product
Product Code. 18662806
  • € 595.00 / 100µL
Description
beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
beta 2-Microglobulin
Monoclonal
Biotin
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient
100 μg
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
567
Store at 4°C in the dark.
IgG2a κ
Flow Cytometry, Immunofluorescence, CyTOF
246-E9.E7 (same as HLA.ABC.m2)
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Primate (Negative)