missing translation for 'onlineSavingsMsg'
Learn More

beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

€ 552.00 - € 595.00

Specifications

Antigen beta 2-Microglobulin
Clone 246-E9.E7 (same as HLA.ABC.m2)
Applications Flow Cytometry, Immunofluorescence, CyTOF
Classification Monoclonal
Conjugate Biotin
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18662806
View Documents
Novus Biologicals
NBP2-47700B-100ul
100 μg
€ 595.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18606565
View Documents
Novus Biologicals
NBP2-47700B
0.1 mL
€ 552.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence, CyTOF
Specifications

Specifications

beta 2-Microglobulin
Flow Cytometry, Immunofluorescence, CyTOF
Biotin
Mouse
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
PBS
567
IgG2a κ
Protein A or G purified
246-E9.E7 (same as HLA.ABC.m2)
Monoclonal
Purified
RUO
Human, Primate (Negative)
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient
Primary
Store at 4°C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.