Learn More
beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47700B
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence, CyTOF
Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| Biotin | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| 567 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, CyTOF | |
| 246-E9.E7 (same as HLA.ABC.m2) | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Primate (Negative) | |
| Purified |