missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-NGF Antibody (5F10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26112-100ul
This item is not returnable.
View return policy
Description
beta-NGF Monoclonal antibody specifically detects beta-NGF in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| beta-NGF | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 5F10 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428 | |
| A synthesized peptide derived from Human beta-NGF [UniProt P01138] | |
| 100 μL | |
| Cancer, Neuroscience | |
| 4803 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction