missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-NGF Antibody (5F10), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 568.00
Specifications
| Antigen | beta-NGF |
|---|---|
| Clone | 5F10 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30036312
|
Novus Biologicals
NBP3-26112-50ul |
50 μL |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30030173
|
Novus Biologicals
NBP3-26112-100ul |
100 μL |
€ 568.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-NGF Monoclonal antibody specifically detects beta-NGF in Human, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| beta-NGF | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428 | |
| A synthesized peptide derived from Human beta-NGF [UniProt P01138] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5F10 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 4803 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title