missing translation for 'onlineSavingsMsg'
Learn More

beta Sarcoglycan Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 213.00 - € 441.00

Specifications

Antigen beta Sarcoglycan
Concentration 1.0 mg/mL
Dilution Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30226359
View Documents
Novus Biologicals
NBP3-38648SS
0.025 mg
€ 213.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30226358
View Documents
Novus Biologicals
NBP3-38648
0.1 mg
€ 441.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta Sarcoglycan Polyclonal antibody specifically detects beta Sarcoglycan in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications

Specifications

beta Sarcoglycan
Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500
Polyclonal
Purified
RUO
Human
43 kDa dystrophin-associated glycoprotein, 43DAG, A3bbeta-SG, beta-sarcoglycan, beta-sarcoglycan(43kD dystrophin-associated glycoprotein), Beta-SG, LGMD2E, limb girdle muscular dystrophy 2E (non-linked families), sarcoglycan, beta (43kD dystrophin-associated glycoprotein), sarcoglycan, beta (43kDa dystrophin-associated glycoprotein), SGC
Partial recombinant protein made to the C-terminal portion of human beta Sarcoglycan [UniProt Q16585].
Primary
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
1.0 mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Signal Transduction
PBS
6443
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.