missing translation for 'onlineSavingsMsg'
Learn More

Beta-Sitosterol (purity>80%), MedChemExpress

Product Code. 16459016
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459016 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459016 Supplier Medchem Express Supplier No. HYN0171
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Beta-Sitosterol (purity>80%) includes β-sitosterol (≥80%), stigmasterol, campesterol and brassicasterol mainly. Beta-Sitosterol is a plant sterol. Beta-Sitosterol (purity>80%) interfere with multiple cell signaling pathways, including cell cycle, apoptosis, proliferation, survival, invasion, angiogenesis, metastasis and inflammation.

Beta-Sitosterol (purity>80%) includes β-sitosterol (≥80%), stigmasterol, campesterol and brassicasterol mainly. Beta-Sitosterol is a plant sterol. Beta-Sitosterol (purity>80%) interfere with multiple cell signaling pathways, including cell cycle, apoptosis, proliferation, survival, invasion, angiogenesis, metastasis and inflammation.

Specifications

Chemical Name or Material Beta-Sitosterol (purity>80%)
CAS 83-46-5
Color White
Purity Grade Notes Research
Molecular Formula C29H50O
Quantity 100 mg
Synonym β-Sitosterol (purity>80%) 22,23-Dihydrostigmasterol (purity>80%)
Solubility Information Ethanol : 3.85 mg/mL (9.28 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.41 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.24 mM; Need ultrasonic)
SMILES CC[C@@H](C(C)C)CC[C@@H](C)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C.[(purity>80%)]
Molecular Weight (g/mol) 414.71
Formula Weight 414.71
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.