missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody (SAP.4G5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB120-11312-0.025ml
This item is not returnable.
View return policy
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| beta I Tubulin | |
| Monoclonal | |
| Unconjugated | |
| beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| No reactivity with other tubulin isotypes is observed. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| SAP.4G5 | |
| Western Blot 1:20000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| TUBB1 | |
| Synthetic peptide corresponding to the C-terminal sequence of beta-tubulin isotype I, coupled to BSA. | |
| 0.025 mL | |
| Cytoskeleton Markers | |
| 81027 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction