missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody (SAP.4G5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 202.65
Specifications
| Antigen | beta I Tubulin |
|---|---|
| Clone | SAP.4G5 |
| Dilution | Western Blot 1:20000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| beta I Tubulin | |
| Western Blot 1:20000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 81027 | |
| Synthetic peptide corresponding to the C-terminal sequence of beta-tubulin isotype I, coupled to BSA. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SAP.4G5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers | |
| beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 | |
| TUBB1 | |
| IgG1 | |
| Unpurified | |
| No reactivity with other tubulin isotypes is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title