missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Binding Buffer DB
Click to view available options
Quantity:
25 mL
1000 mL
Unit Size:
1000mL
25mL
Description
NucleoSpin™ gDNA Clean-Up Kit is used for post clean-up and concentration of large-sized DNA.
- Effective post clean-up and concentration of large DNA fragments (e.g., pre-purified DNA, DNA from enzymatic reactions).
- Typical downstream applications: qPCR, enzymatic reactions, Southern blotting, STR amplification.
- Not recommended for gel or PCR clean-up.
- High recovery rates up to 90% for DNA fragments 100 bp–50 kbp.
- Concentrate DNA sample easily and faster than with microdialysis filtrations units.
- 15 min./10 preps – time-saving procedure.
Specifications
Specifications
| Chemical Name or Material | Guanidine hydrochloride |
| CAS | 50-01-1 |
| Color | Colorless |
| For Use With (Application) | Post clean-up and concentration of large sized DNA |
| For Use With (Equipment) | NucleoSpin gDNA Clean-up |
| Physical Form | Liquid |
| Quantity | 1000 mL |
| Odor | Alcoholic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction