missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated Human IgG Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech BIOTIN-10284-100UL
This item is not returnable.
View return policy
Description
The Human IgG antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of Human IgG. This antibody recognizes human antigen. The Human IgG antibody has been validated for the following applications: IHC analysis. This antibody is biotin.Specifications
| IgG | |
| Polyclonal | |
| Biotin | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunohistochemistry | |
| 1000 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| Human | |
| Conjugated Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction