missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin-conjugated Human IgG Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 140.00 - € 351.27
Specifications
| Antigen | IgG |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237572
|
Proteintech
BIOTIN-10284-100UL |
100 μL |
€ 351.27
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234261
|
Proteintech
Biotin-10284-20UL |
20 μL |
€ 140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Human IgG antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of Human IgG. This antibody recognizes human antigen. The Human IgG antibody has been validated for the following applications: IHC analysis. This antibody is biotin.Specifications
| IgG | |
| Immunohistochemistry | |
| Biotin | |
| Rabbit | |
| Human | |
| IgG | |
| Antigen affinity purification | |
| Conjugated Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title