Learn More
Fisherbrand™ Black Phenolic Caps
Product Code. 16498115 Shop All Fisherbrand Products
Change view
Click to view available options
Neck Finish:
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
Quantity:
12
576
1600
3100
5500
Unit Size:
Pack of 1600
Pack of 3100
Pack of 5500
Pack of 576
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Quantity unitSize
16498115 20-400 5500 Pack of 5500
16174397 38-400 1600 Pack of 1600
16134587 20-400 576 Pack of 576
16144587 24-400 12 Pack of 576
16154587 22-400 12 Pack of 576
16174587 18-400 12 Pack of 576
16164587 15-425 576 Pack of 576
16184587 28-400 576 Pack of 576
16134617 28-400 3100 Pack of 3100
16104597 38-400 12 Pack of 576
16194587 33-400 576 Pack of 576
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 16498115
Only null left
Add to basket
Add to basket
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Specifications
Color Black
Quantity 5500
Closure Color Black
Neck Finish 20-400
Closure Included No
Closure Liner Polyethylene Cone
Closure Material Phenolic