Learn More
BMPR1B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26696-1-AP-150UL
Description
The BMPR1B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BMPR1B. This antibody recognizes human, mouse, rat antigen. The BMPR1B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BMPR1B | |
| 750 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ALK 6, ALK6, BMP type 1B receptor, BMPR 1B, BMPR1B, CDw293 | |
| 150 μL | |
| 658 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |