Learn More
BMPR1B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26696-1-AP-150UL
 View more versions of this product
Product Code. 18075485
  • € 410.00 / 150µL
Description
The BMPR1B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BMPR1B. This antibody recognizes human, mouse, rat antigen. The BMPR1B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
BMPR1B
750 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
ALK 6, ALK6, BMP type 1B receptor, BMPR 1B, BMPR1B, CDw293
150 μL
658
Store at -20°C. Stable for one year after shipment.