missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMPR1B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | BMPR1B |
|---|---|
| Concentration | 750 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18075485
|
Proteintech
26696-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18008745
|
Proteintech
26696-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BMPR1B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BMPR1B. This antibody recognizes human, mouse, rat antigen. The BMPR1B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BMPR1B | |
| Western Blot, ELISA | |
| Rabbit | |
| 658 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 750 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| ALK 6, ALK6, BMP type 1B receptor, BMPR 1B, BMPR1B, CDw293 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title