missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BMS 191011

Product Code. 18055282
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18055282

Brand: Tocris Bioscience™ 2665/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent BKCa (KCa1.1) channel opener

Potent BKCa channel opener (large-conductance Ca2+-activated potassium channel, KCa1.1). Neuroprotectant in two distinct animal models of stroke (MCAO in the SHR rat and a normotensive model of focal stroke).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202821-81-6
Molecular Formula C16H10ClF3N2O3

Specifications

Quantity 10 mg
Formula Weight 370.71
Purity 98%
Molecular Weight (g/mol) 370.712
Chemical Name or Material BMS 191011
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.