Learn More
Witeg™ Iodine Determination Flask
Witeg™ Iodine Determination Flask is an Erlenmeyer-style flask with a ground glass joint, a modified stopper, and a funnel-shaped top.
Brand:  Witeg™ 2 438 250
Additional Details : Weight : 0.17000kg
 View more versions of this product
Product Code. 10037281
  • € 72.55 / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
  • Stopper handle projects above the liquid seal trough to facilitate removal
  • Ground glass joint prevents vapors from escaping, improving the accuracy of your analysis
  • Funnel-shaped top to ensure material entry and minimize spillage
Specifications
Specifications
250 mL
Borosilicate Glass (3.3)
Hollow Stopper with Long Stem