Learn More
Witeg™ Iodine Determination Flask
Witeg™ Iodine Determination Flask is an Erlenmeyer-style flask with a ground glass joint, a modified stopper, and a funnel-shaped top.
Brand: Witeg™ 2 438 500
Additional Details : Weight : 0.21400kg
Description
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion
- Stopper handle projects above the liquid seal trough to facilitate removal
- Ground glass joint prevents vapors from escaping, improving the accuracy of your analysis
- Funnel-shaped top to ensure material entry and minimize spillage
Specifications
500 mL | |
Borosilicate Glass (3.3) | |
Hollow Stopper with Long Stem |