Learn More
Kimble Chase™ Borosilicate Glass Serum Vials without Closures
Click to view available options
Volume (Metric):
2 mL
5 mL
Unit Size:
Pack of 4640
Pack of 864
Description
- Special design provides extra strength for freeze-drying applications
- Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
- Special blowback design provides ease of use for lyophillization stoppers
- Autoclavable
- Aluminum seals, stoppers and septa are available separately
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Diameter (Metric) Outer | 23 mm |
| Material | Borosilicate Glass (Type 1 Class A) |
| Volume (Metric) | 5 mL |
| No. per Case | 864 |
| Height (Metric) | 47 mm |
| Closure Size | A/S 20 |