Learn More
Brady™ BradyJet™ J7300 Color Label Printer
Product Code. 30058104
Change view
Click to view available options
Model:
J7300 EU
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
30058104 J7300 EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30058104 Supplier Brady™ Supplier No. 176883
Only null left
Add to basket
Add to basket
Unleash your true colors with the BradyJet™ J7300 Color Label Printer. Enjoy fast, industrial-grade full color for variable print jobs from the production line to the lab.
  • Ideal for industrial labelling that changes frequently, from regulatory signage and Lean visuals to barcoding, lab ID, GHS labelling and UL/component ID
  • LabelSense™ technology enables printer, materials and inks to talk to each other for automatic part recognition, setup and adjustments
  • Dual CMY and true black (K) ink cartridges deliver vibrant colors, deep black, dark tones, readable barcodes and sharp text job after job
  • Prints smear-free inks on labels that resist water, chemicals and abrasions for up to two-year outdoor durability (even longer indoors)
  • Print Utility dashboard helps you plan, budget and troubleshoot with status alerts, a job cost calculator, remote monitoring and more
  • Exterior LED lighting and clear cover communicate real-time printer status
  • Ships with the free Brady Workstation Basic software that includes fundamental apps and features to create labels.
  • Optimized Windows™ driver lets you connect to third-party software and print PDFs
Specifications
Product Type Printer
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Inkjet
Length (English) 9.09 in.
Width (English) 14.81 in.
Length (Metric) 230.89 mm
Width (Metric) 376.30 mm
Depth (English) 20.303 in.
Depth (Metric) 515.70 mm
Color Capability Full Color
Description BradyJet J7300 Color Label Printer EU
Display Size 155.44 x 91.95 mm
Includes BradyJet J7300 Printer, CMY Ink Cartridge with Built-in Printhead, a Black Ink Cartridge with Built-in Printhead, a Brady Workstation Basic Software, a J-Align Roll, a Media Starter B-2595 Vinyl Roll, an AC Adapter, a Power Cord, a USB Cable and a USB Flash Drive with Product Information.
Languages Bahasa (Indonesian), Bahasa Malay, Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French, German, Greek, Hungarian, Italian, Japanese, Korean, Norwegian (Bokmål), Polish, Portuguese, Punjabi, Slovak, Slovenian, Spanish (universal), Swedish, Thai, Turkish, Vietnamese
Memory 32 GB, expandable through USB
Model J7300 EU
Print Speed 30.48 mm/s
Resolution 4800 dpi
Software Type Brady LINK260;Brady Workstation;Windows based driver for 3rd party software use
Connector Types Cabled, Ethernet, USB-C, USB Host
Operating System Windows 10
Weight (English) 36 lb
Weight (Metric) 16.33 kg
Certifications/Compliance CE, C-UL US, FCC Class A, RoHS directive 2011/65/EU, UL Listed for US
For Use With (Application) Asset tracking, Barcode labelling, Component and equipment labelling, Exit and egress marking, Facility identification, General identification, Hazard communication identification, Healthcare and Hospital labelling, Inspection tagging, Inventory and inspection labelling, Laboratory labelling, Lean and 5S labelling, Lockout tagout, Marine labelling, Pipe and valve marking, Safety identification, Warehouse marking
Quantity 1
Warranty 2 Years
Amperage 100/240 V
Power Supply Details AC Adapter
Voltage 100/240 V
Keyboard Type None