missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ BrassTec 947 Gloves

Product Code. 15818004
Change view
Click to view available options
Size: 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15818004 Size: 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15818004 Supplier Honeywell™ Supplier No. 947T10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell™ BrassTec 947 Gloves offer heat protection up to a contact heat of 250°C, while providing excellent grip.

  • Special silicone coating with very high temperature resistance good cut resistance multi-layer system contact heat up to 250°C
  • Special coated palm for excellent grip molten metal beads and rolls off good thermal insulation radiant heat up to 1,000°C conical shape allows easy donning and doffing moisture-repellent
TRUSTED_SUSTAINABILITY

Specifications

Description 3 -layer-system : para-aramid, cotton and Aluminum on the back. Silicone on palm
Color Silver, Gray
Size 10
Temperature (Metric) Max 250°C
Thickness (Metric) 3.1 mm
Length (Metric) 29 to 31 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.