Learn More
BST2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-13560-20UL
 View more versions of this product
Product Code. 18027895
  • € 140.00 / 20µL
Description
The BST2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BST2. This antibody recognizes human, mouse antigen. The BST2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
BST2
1000 μg/mL
Bone marrow stromal antigen 2, BST 2, BST2, CD317, HM1.24 antigen, Tetherin
20 μL
684
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse
150 kDa