missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BST2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 140.00 - € 450.00
Specifications
| Antigen | BST2 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18004275
|
Proteintech
CL488-13560-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18027895
|
Proteintech
CL488-13560-20UL |
20 μL |
€ 140.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BST2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BST2. This antibody recognizes human, mouse antigen. The BST2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| BST2 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse | |
| Bone marrow stromal antigen 2, BST 2, BST2, CD317, HM1.24 antigen, Tetherin | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 684 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title