missing translation for 'onlineSavingsMsg'
Learn More

Budesonide, MedChemExpress

Product Code. 16481086
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16481086

Brand: Medchem Express HY13580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Budesonide, an inhaled glucocortical steroid, is an orally active glucocorticoid receptor agonist. Budesonide decreases the size of lung tumors, reverses DNA hypomethylation and modulates mRNA expression of genes. Budesonide is an anti-inflammatory agent used for asthma.

Budesonide, an inhaled glucocortical steroid, is an orally active glucocorticoid receptor agonist. Budesonide decreases the size of lung tumors, reverses DNA hypomethylation and modulates mRNA expression of genes. Budesonide is an anti-inflammatory agent used for asthma.

Specifications

Chemical Name or Material Budesonide
CAS 51333-22-3
Color White
Purity Grade Notes Research
Molecular Formula C25H34O6
Quantity 200 mg
Solubility Information DMSO : 215 mg/mL (499.38 mM; Need ultrasonic and warming)
SMILES O=C(C=C[C@@]12C)C=C1CC[C@@]([C@]2([H])[C@@H](O)C[C@@]34C)([H])[C@]3([H])C[C@]5([H])[C@@]4(C(CO)=O)OC(CCC)O5
Molecular Weight (g/mol) 430.53
Formula Weight 430.53
Percent Purity 98.33%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.