missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BX 513 hydrochloride
GREENER_CHOICE

Product Code. 18005292
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18005292

Brand: Tocris Bioscience™ 2769/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective CCR1 antagonist

Selective CCR1 receptor antagonist (Ki values are 0.04, > 10, > 10 and > 10 nM for CCR1, CCR5, CXCR2 and CXCR4 receptors respectively). Inhibits MIP-1α-induced intracellular calcium mobilization (IC50 = 2.5 μM). Also a full inverse agonist at US28, a HCMV-encoded chemokine receptor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216540-18-9
Molecular Formula C28H30Cl2N2O

Specifications

Quantity 50 mg
Formula Weight 481.46
Purity >99%
Molecular Weight (g/mol) 481.461
Chemical Name or Material BX 513 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.